Learn More
Diapharma Group, Inc. Chromogenix S-2765, 25 mg
SDP
Catalog No. NC9781310
Encompass
Chromogenix S-2765 is a chromogenic substrate for determination of Factor Xa activity. It is also very sensitive to trypsin. S-2765 is suitable for measuring FXa inhibition in heparin anti-Xa assays and antithrombin anti-Xa assays.
Catalog No. NC9781310 Supplier Diapharma Group, Inc. Supplier No. S821413
Chromogenix S-2765 is a chromogenic substrate for determination of Factor Xa activity. It is also very sensitive to trypsin. S-2765 is suitable for measuring FXa inhibition in heparin anti-Xa assays and antithrombin anti-Xa assays.