Learn More
Diapharma Group, Inc. Chromogenix S-2251™, Chromogenic Substrate For Plasmin Detection, Reliable For Fibrinolysis Studies, 25 mg
SDP
Catalog No. NC9492015
Encompass
Chromogenix S-2251 is a specialized chromogenic substrate designed for the accurate detection and quantification of plasmin and streptokinase-activated plasminogen in plasma. It provides reliable results for hemostasis and fibrinolysis studies, making it essential for researchers in these fields. Its unique chromogenic method enhances detection, ensuring elevated reproducibility and precision in assays. This product is determined by its high-quality formulation and extensive application range.
Catalog No. NC9492015 Supplier Diapharma Group, Inc. Supplier No. S820332
Chromogenix S-2251 is a specialized chromogenic substrate designed for the accurate detection and quantification of plasmin and streptokinase-activated plasminogen in plasma. It provides reliable results for hemostasis and fibrinolysis studies, making it essential for researchers in these fields. Its unique chromogenic method enhances detection, ensuring elevated reproducibility and precision in assays. This product is determined by its high-quality formulation and extensive application range.