Learn More
Enzo Life Sciences Chromogranin A (human) monoclonal antibody (PHE5) (Ready-to-Use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ30932
Encompass_Preferred
Clone: PHE5. Isotype: IgG1. Immunogen: Human pheochromocytoma. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: PHE5 is specific to chromogranin A, a highly acidic protein associated with the “dense core” granules of many cells of the neural and neuroendocrine system. It reacts with neuroendocrine cells of the gut, pancreatic islet and adrenal medulla, but does not react with any cells in the brain or with melanoma cells. The antibody stains carcinoid tumors, pheochromocytomas and paragangliomas. It is nonreactive with carcinomas, lymphomas and sarcomas. Applications: IHC (PS). Purity: Affinity purified. Formulation: Liquid. Ready-to-use solution in PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-2485