Learn More
Biogenex Laboratories Chromogranin A : LK2H10 : 1mL
SDP
Supplier:  Biogenex Laboratories MU126UC
Encompass_Preferred
This antibody recognizes Chromogranin A (68kD) and other related chromogranin polypeptides from human, monkey, and pig. Chromogranin is widely distributed and through immunohistochemistry, chromogranin has been demonstrated in several elements of the diffuse neuroendocrine system, including anterior pituitary, thyroid parafollicular C cells, parathyroid chief cells, pancreatic islet cells, intestinal enteroendocrine cells, and tumors derived from these cells. The measurement of Chromogranin A has become a valuable tool in the investigation of neuroendocrine neoplasia.
Catalog No. 50-197-8877