Learn More
Enzo Life Sciences Chromogranin A polyclonal antibody (100µl)
SDP
Catalog No. 502012813
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1-15 (L1PVNSPMNKGDTEVM15) of native human chromogranin A, conjugated to KLH. Species reactivity: Human. Applications: ELISA, IHC. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2813 Supplier Enzo Life Sciences Supplier No. ENZABS2680100
Immunogen: Synthetic peptide corresponding to aa 1-15 (L1PVNSPMNKGDTEVM15) of native human chromogranin A, conjugated to KLH. Species reactivity: Human. Applications: ELISA, IHC. Formulation: Liquid. In PBS containing 0.09% sodium azide.