Learn More
AdipoGen Chromoionophore VI
SDP
Catalog No. 502561037
Encompass_Preferred
Chemical. CAS 138833-47-3. Formula C38H46Br2O5. MW BD9837. Chromoionophore VI ETH 7075, a synthetic compound composed of a chromophore and an ionophore is as an acidic lipophilic pH indicator and is generally used during the fabrication of optical sensors/membranes. Chromoionophore VI has been designed to selectively interact with transition metal ions, such as copper, nickel, and zinc, and to produce a fluorescent signal in response to their presence. This property makes Chromoionophore VI an attractive tool for studying the processes by which these metals are transported and utilized in biological systems and a wide variety of biological processes, including apoptosis, oxidative stress, and metal ion homeostasis. Chromoionophore VI is a powerful tool for studying the interactions between ions and molecules, as well as for studying the effects of ionic strength on biochemical and physiological processes.
Catalog No. 50-256-1037 Supplier AdipoGen Supplier No. CDXD0810M100
Chemical. CAS 138833-47-3. Formula C38H46Br2O5. MW BD9837. Chromoionophore VI ETH 7075, a synthetic compound composed of a chromophore and an ionophore is as an acidic lipophilic pH indicator and is generally used during the fabrication of optical sensors/membranes. Chromoionophore VI has been designed to selectively interact with transition metal ions, such as copper, nickel, and zinc, and to produce a fluorescent signal in response to their presence. This property makes Chromoionophore VI an attractive tool for studying the processes by which these metals are transported and utilized in biological systems and a wide variety of biological processes, including apoptosis, oxidative stress, and metal ion homeostasis. Chromoionophore VI is a powerful tool for studying the interactions between ions and molecules, as well as for studying the effects of ionic strength on biochemical and physiological processes.