Learn More
MilliporeSigma™ Supelco™ Chromolith™ Guard Cartridge Holder
Catalog No. 111023074 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Line:
Chromolith
Catalog No. Product Line
11-102-3074 Chromolith
1 options
Catalog No. 11-102-3074 Supplier MilliporeSigma™ Supelco™ Supplier No. 1523550001
Add to Cart
Add to Cart
The Chromolith™ 5-2 guard cartridge holder bioinert is suitable to connect any Chromolith™ guard cartridge to an analytical Chromolith™ HPLC column.
  • The Chromolith™ 5-2 guard cartridge holder bioinert is pressure stable up to 400 bar and made of PEEK-lined Stainless Steel.
  • The holder is suitable to connect any Chromolith™ guard cartridge of 5 mm length and 2 mm I.D. to an analytical Chromolith™ HPLC column.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC
Max. Pressure 400 bar
Product Line Chromolith
Type Cartridge Holder
Material PEEK-Lined Stainless Steel