Learn More
Cayman Chemical Chromomycin A3, Purity: ≥95%, CAS: 7059-24-7, 10 mg
SDP
Catalog No. 502955705
Encompass_Preferred
A fluorescent probe and antitumor agent; displays ex/em of 445/575 nm; specifically binds GC base pairs on DNA; can be used to characterize heterochromatin in plants; cytotoxic against non-small cell lung cancer and cervical cancer in vitro (IC50s = 1, 42, 60, and 40 nM for HCC44, A549, ME180, and HeLa cells, respectively); inhibits oxidative stress- and DNA damage-induced neuronal injury
Catalog No. 50-295-5705 Supplier Cayman Chemical Supplier No. 1131510 MG
A fluorescent probe and antitumor agent; displays ex/em of 445/575 nm; specifically binds GC base pairs on DNA; can be used to characterize heterochromatin in plants; cytotoxic against non-small cell lung cancer and cervical cancer in vitro (IC50s = 1, 42, 60, and 40 nM for HCC44, A549, ME180, and HeLa cells, respectively); inhibits oxidative stress- and DNA damage-induced neuronal injury