Learn More
Medchemexpress LLC HY-W040129 5mg Medchemexpress, Chromomycin A3 CAS:7059-24-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0401295MG
Encompass_Preferred
Medchemexpress, HY-W040129 5mg Chromomycin A3 CAS:7059-24-7 Chromomycin A3 is an aureolic acid-type antitumor antibiotic. Chromomycin A3 forms dimeric complexes with divalent cations, such as Mg2+, which strongly binds to the GC rich sequence of DNA to inhibit DNA replication and transcription. Chromomycin A3 has a variety of utilities as a staining agent for human sperm chromatin, autophagy inducing agent, and apoptosis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0741