Learn More
Abcam Chromomycin A3, fluorescent DNA stain, 5MG
Catalog No. 502706200
Encompass_Preferred
MW 1183.2 Da, Purity >98%. Anthraquinone antibiotic, fluorescent DNA stain. Binds transcription factors Sp1 and SP3 to DNA to inhibit RNA synthesis. Shows anti-bacterial, anti-fungal and antitumor activities. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6200 Supplier Abcam Supplier No. AB1427185MG
MW 1183.2 Da, Purity >98%. Anthraquinone antibiotic, fluorescent DNA stain. Binds transcription factors Sp1 and SP3 to DNA to inhibit RNA synthesis. Shows anti-bacterial, anti-fungal and antitumor activities. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement