Learn More
Leica Biosystems BOND ChromoPlex II Dual Detection Kit, 2-plex detection system, immunohistochemistry (IHC), chromogenic in situ hybridization (CISH), biotin-free, mouse and rabbit antibody compatible, 4.5 hours (BOND-III), 6 hours (BOND-Max)
Catalog No. NC2747668
Encompass
BOND ChromoPlex II Dual Detection Kit, 2-plex detection system, immunohistochemistry (IHC), chromogenic in situ hybridization (CISH), biotin-free, mouse and rabbit antibody compatible, 4.5 hours (BOND-III), 6 hours (BOND-Max)
Catalog No. NC2747668 Supplier Leica Biosystems Supplier No. DS9494
Add to Cart
Add to Cart
Request a Quote
BOND ChromoPlex II Dual Detection Kit, 2-plex detection system, immunohistochemistry (IHC), chromogenic in situ hybridization (CISH), biotin-free, mouse and rabbit antibody compatible, 4.5 hours (BOND-III), 6 hours (BOND-Max)