Learn More
Empire Genomics Chromosome 17 Control FISH Probe
Catalog No. NC3725960
Encompass
Empire Genomics' Chromosome 17 Control FISH Probe is designed to hybridize near the centromeric region of the chromosome. Control probes are useful for both chromosome enumeration and an internal control used along side one of our gene specific probes.
Catalog No. NC3725960 Supplier Empire Genomics Supplier No. CHR1710GR
Empire Genomics' Chromosome 17 Control FISH Probe is designed to hybridize near the centromeric region of the chromosome. Control probes are useful for both chromosome enumeration and an internal control used along side one of our gene specific probes.