Learn More
CPC Scientific H-Ser-Asp-Glu-Asp-Ser-Asp-Gly-Asp-Arg-Pro-Gln-Ala-Ser-Pro-Gly-Leu-Gly-Pro-Gly-Pro-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PANC001A
Encompass
Chromostatin-20 corresponding to the first 20 N-terminal amino acids of bovine chromogranin A that produces a dose-dependent inhibition of catecholamine secretion from cultured chromaffin cells in the range of 10-9 to 10-6 M (ID50 approx. 5nM). It also modulates endocrine of catecholamine-associated response. ONE-LETTER SEQUENCE: SDEDSDGDRPQASPGLGPGPMOLECULAR FORMULA: C78H120N24O35MOLECULAR WEIGHT:1954STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [133633-11-1], RESEARCH AREA: DiabetesREFERENCES: E. Galindo et al., Proc. Nat. Acad. Sci. USA, 88, 1426 (1991)
Catalog No. 50-211-0072