Learn More
Chromotrope 2R, Sigma-Aldrich
Catalog No. 01259027 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
01-259-027 100 g
1 options
Catalog No. 01-259-027 Supplier MilliporeSigma Supelco Supplier No. C3143100G
Add to Cart
Add to Cart
Powder stain for modified Gomori trichrome staining, ideal for histological staining of eosinophils and microsporidia.
Chromotrope 2R has been used in the Gomori trichrome staining in osteochondral tissues. It has also been used for the staining of eosinophils in lung sections.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Type Stain Kit