Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromotropic acid disodium dihydrate | 5808-22-0 | MFCD00150612 | >98.0% | 400.29 | C10H10Na2O10S2 | 5G
SDP

Catalog No. 5000235534
Encompass_Preferred

Chromotropic acid disodium dihydrate is a solid, disodium salt reagent used for colorimetric and analytical detection of formaldehyde and related assays. It is supplied in laboratory quantities for analytical applications and is documented with a datasheet and safety data sheet.

  • Used for colorimetric detection of formaldehyde.
  • Molecular formula C10H10Na2O10S2.
  • Molecular weight 400.29 g/mol.
  • Solid form, light brown to brown appearance.
  • Available in small lab pack sizes (for example, 5 g and larger).
  • Accompanied by datasheet and safety data sheet documentation.

Catalog No. 50-002-35534 Supplier Medchemexpress LLC Supplier No. HY1230655G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromotropic acid disodium dihydrate is a solid, disodium salt reagent used for colorimetric and analytical detection of formaldehyde and related assays. It is supplied in laboratory quantities for analytical applications and is documented with a datasheet and safety data sheet.

  • Used for colorimetric detection of formaldehyde.
  • Molecular formula C10H10Na2O10S2.
  • Molecular weight 400.29 g/mol.
  • Solid form, light brown to brown appearance.
  • Available in small lab pack sizes (for example, 5 g and larger).
  • Accompanied by datasheet and safety data sheet documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.