Learn More
Medchemexpress LLC Chromotropic acid disodium dihydrate | 5808-22-0 | MFCD00150612 | 400.29 g/mol | C10H10Na2O10S2 | 25G
SDP
Catalog No. 5000235747
Encompass_Preferred
Chromotropic acid disodium dihydrate is the disodium salt dihydrate of chromotropic acid used as an analytical reagent for colorimetric detection of formaldehyde and as a staining/dyeing reagent. It is supplied in multiple pack sizes, including a 25 g unit, and is characterized by the molecular formula C10H10Na2O10S2 and a molecular weight of 400.29 g/mol.
  • Reagent for colorimetric detection of formaldehyde.
  • Available in multiple pack sizes, including 25 g.
  • Provided as the disodium salt dihydrate form.
  • Molecular formula C10H10Na2O10S2; molecular weight 400.29 g/mol.
  • Suitable for analytical and staining applications.
Catalog No. 50-002-35747 Supplier Medchemexpress LLC Supplier No. HY12306525G
Chromotropic acid disodium dihydrate is the disodium salt dihydrate of chromotropic acid used as an analytical reagent for colorimetric detection of formaldehyde and as a staining/dyeing reagent. It is supplied in multiple pack sizes, including a 25 g unit, and is characterized by the molecular formula C10H10Na2O10S2 and a molecular weight of 400.29 g/mol.
  • Reagent for colorimetric detection of formaldehyde.
  • Available in multiple pack sizes, including 25 g.
  • Provided as the disodium salt dihydrate form.
  • Molecular formula C10H10Na2O10S2; molecular weight 400.29 g/mol.
  • Suitable for analytical and staining applications.