Learn More
Medchemexpress LLC Chromotropic acid disodium dihydrate | 5808-22-0 | MFCD00150612 | 98.5% | 400.29 g/mol | C10H10Na2O10S2 | 500G
SDP
Catalog No. 5000238795
Encompass_Preferred
Chromotropic acid disodium dihydrate is the disodium salt dihydrate of chromotropic acid used as a reagent and dye in analytical and biochemical applications. It is commonly applied for colorimetric detection of formaldehyde and as an indicator or staining agent. The compound is a water-soluble crystalline powder with molecular formula C10H10Na2O10S2 and molecular weight 400.29 g/mol, and is available in laboratory pack sizes up to 500 g.
Catalog No. 50-002-38795 Supplier Medchemexpress LLC Supplier No. HY123065500G
Chromotropic acid disodium dihydrate is the disodium salt dihydrate of chromotropic acid used as a reagent and dye in analytical and biochemical applications. It is commonly applied for colorimetric detection of formaldehyde and as an indicator or staining agent. The compound is a water-soluble crystalline powder with molecular formula C10H10Na2O10S2 and molecular weight 400.29 g/mol, and is available in laboratory pack sizes up to 500 g.