Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromotropic acid disodium dihydrate | 5808-22-0 | MFCD00150612 | 400.29 g/mol | C10H10Na2O10S2 | 50G
SDP

Catalog No. 5000237580
Encompass_Preferred

Chromotropic acid disodium dihydrate is a solid laboratory reagent used primarily for the colorimetric detection of formaldehyde. The disodium salt dihydrate form is formulated for analytical and research use and dissolves in water with heating and ultrasonic assistance, enabling preparation of aqueous assay solutions.

  • Can be used for the detection of formaldehyde.
  • Soluble in water (≈50 mg/mL) with ultrasonic and warming.
  • Molecular weight 400.29 g/mol.
  • Available in laboratory-scale pack sizes for analytical use.

Catalog No. 50-002-37580 Supplier Medchemexpress LLC Supplier No. HY12306550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromotropic acid disodium dihydrate is a solid laboratory reagent used primarily for the colorimetric detection of formaldehyde. The disodium salt dihydrate form is formulated for analytical and research use and dissolves in water with heating and ultrasonic assistance, enabling preparation of aqueous assay solutions.

  • Can be used for the detection of formaldehyde.
  • Soluble in water (≈50 mg/mL) with ultrasonic and warming.
  • Molecular weight 400.29 g/mol.
  • Available in laboratory-scale pack sizes for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.