Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromotropic Acid, Disodium Dihydrate, Reagents

Catalog No. 01337312
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-312 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-312 Supplier Reagents Holdings Llc Supplier No. C217970025C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromotropic Acid, Disodium Dihydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 5808-22-0
Color White to Off-White
Packaging Glass Bottle
Molecular Formula C10H10Na2O10S2
MDL Number MFCD00150612
Quantity 25 g
Specific Gravity 1.52
Synonym 4,5-dihydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt Dihydrate
InChI Key QUEAKWJKJBFNEG-UHFFFAOYSA-L
SMILES O.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]
IUPAC Name 2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
Molecular Weight (g/mol) 400.28
Grade Pure
Physical Form Solid
Chemical Name or Material Chromotropic Acid, Disodium Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.