Learn More
Medchemexpress LLC Chromotropic acid disodium salt dihydrate | 5808-22-0 | MFCD00150612 | 400.29 g/mol | C10H6Na2O8S2·2H2O | 100G
SDP
Catalog No. 5000237340
Encompass_Preferred
Chromotropic acid disodium salt dihydrate is the disodium salt form of chromotropic acid used as a chromogenic reagent for qualitative and quantitative detection of formaldehyde. It is supplied as a solid reagent (chemical formula C10H6Na2O8S2·2H2O; molecular weight 400.29 g/mol) for analytical and reagent-preparation applications.
  • Used for colorimetric detection of formaldehyde.
  • Solid reagent suitable for reagent preparation and analytical assays.
  • Molecular weight 400.29 g/mol; chemical formula C10H6Na2O8S2·2H2O.
  • Available in common laboratory pack sizes, including 100 g.
  • Store dry and cool; in solution, store at -80°C for long-term or -20°C for shorter-term storage.
Catalog No. 50-002-37340 Supplier Medchemexpress LLC Supplier No. HY123065100G
Chromotropic acid disodium salt dihydrate is the disodium salt form of chromotropic acid used as a chromogenic reagent for qualitative and quantitative detection of formaldehyde. It is supplied as a solid reagent (chemical formula C10H6Na2O8S2·2H2O; molecular weight 400.29 g/mol) for analytical and reagent-preparation applications.
  • Used for colorimetric detection of formaldehyde.
  • Solid reagent suitable for reagent preparation and analytical assays.
  • Molecular weight 400.29 g/mol; chemical formula C10H6Na2O8S2·2H2O.
  • Available in common laboratory pack sizes, including 100 g.
  • Store dry and cool; in solution, store at -80°C for long-term or -20°C for shorter-term storage.