Learn More
CPC Scientific N-Methylsulfonyl-DPhe-Gly-Arg-pNA (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110480
Encompass
Determination of t-PA(tissue plasminogen activator) in purified preparation and in cell culture supernatents in the micro gram scale. Working concentration ~0.25 mM.ONE-LETTER SEQUENCE: N-Methylsulfonyl-DPhe-GR-pNAMOLECULAR FORMULA: C24H32N8O7S1MOLECULAR WEIGHT:576.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Apoptosis
Catalog No. 50-211-0480 Supplier CPC Scientific Supplier No. SUBS011A
Determination of t-PA(tissue plasminogen activator) in purified preparation and in cell culture supernatents in the micro gram scale. Working concentration ~0.25 mM.ONE-LETTER SEQUENCE: N-Methylsulfonyl-DPhe-GR-pNAMOLECULAR FORMULA: C24H32N8O7S1MOLECULAR WEIGHT:576.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Apoptosis