Learn More
Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements
Shop All Fisherbrand ProductsDescription
Chromplete Isopropanol meets the analytical requirements of USP and is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- Pesticide Analysis by GC or HPLC
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Specifications
Specifications
| Melting Point | -90°C |
| Absorbance | ≤ 1.0, ≤ 0.20, ≤ 0.10, ≤ 0.015 |
| Color | ≤ 5 APHA |
| CAS Min % | 0.999 |
| Boiling Point | 83°C |
| Quantity | 4 L |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | ≤ 1.0 ppm |
| UN Number | 1219 |
| Show More |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 3) |
| Hazard Statement | Causes serious eye irritation, Highly flammable liquid and vapor, May cause drowsiness or dizziness |
| ShelfLife | 1825 days |
| EINECSNumber | 200-661-7 |
| RTECSNumber | NT8050000 |
| storageNote1 | At room temperature in sealed containers |
| Recommended Storage | Room temperature flammable storage |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.