Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Water, HPLC/GC, and Spectrometry

Catalog No. T001034000 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T001034000 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T001034000

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Water is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry. | CAS: 7732-18-5 | H2O | 18.02 g/mol

Chromplete Water is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name water
SMILES O

Specifications

Absorbance ≤ 0.01, ≤ 0.005
pH 5.5-7.0
Color ≤5 APHA
CAS Min % 0.99
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.01
Grade Chromplete
Packaging Amber Glass
Residue after Evaporation ≤ 1.0 ppm
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
ShelfLife 1825 Days
EINECSNumber 231-791-2
RTECSNumber ZC0110000
storageNote1 At room temperature in sealed containers
Recommended Storage Room Temperature

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.