Learn More
Chrom Tech Inc Chiral-AGP, 100 x 4.0mm, 5um, Column
SDP
Catalog No. NC9986286
Encompass
CHIRALPAK AGP is the second generation chiral separation column based on the use of α1-acid glycoprotein (AGP) as the CSP. Through a patented process α1-AGP has been immobilized on porous, spherical silica particles (5μm). The surface chemistry of the silica proves a stable chiral separation material with extremely broad applicability.
Catalog No. NC9986286 Supplier Chrom Tech Inc Supplier No. CT20104
CHIRALPAK AGP is the second generation chiral separation column based on the use of α1-acid glycoprotein (AGP) as the CSP. Through a patented process α1-AGP has been immobilized on porous, spherical silica particles (5μm). The surface chemistry of the silica proves a stable chiral separation material with extremely broad applicability.