Learn More
Alkali Scientific ChromX™ Orientation Chromogenic Agar, 20 mL, 10/Pk
SDP
Catalog No. 5000662685
Encompass_Preferred
ChromX™ Chromogenic Culture Media are specialized selective and differential agar formulations designed for rapid isolation and presumptive identification of clinically relevant microorganisms. The chromogenic substrates incorporated into the medium produce distinct colony colors when metabolized by target organisms, allowing laboratories to visually differentiate species directly on the plate. This capability reduces additional confirmation testing and improves diagnostic efficiency in routine microbiology workflows. Each plate is supplied ready to use with approximately 20 mL of prepared medium, supporting consistent growth conditions and reliable performance. Packaged in convenient multi plate formats, ChromX™ media are ideal for clinical laboratories, food safety testing, environmental monitoring, and research facilities requiring accurate microbial detection and streamlined laboratory processing.
Catalog No. 50-006-62685 Supplier Alkali Scientific Supplier No. CHROMX18
ChromX™ Chromogenic Culture Media are specialized selective and differential agar formulations designed for rapid isolation and presumptive identification of clinically relevant microorganisms. The chromogenic substrates incorporated into the medium produce distinct colony colors when metabolized by target organisms, allowing laboratories to visually differentiate species directly on the plate. This capability reduces additional confirmation testing and improves diagnostic efficiency in routine microbiology workflows. Each plate is supplied ready to use with approximately 20 mL of prepared medium, supporting consistent growth conditions and reliable performance. Packaged in convenient multi plate formats, ChromX™ media are ideal for clinical laboratories, food safety testing, environmental monitoring, and research facilities requiring accurate microbial detection and streamlined laboratory processing.