Learn More
Chrono Log Corporation CHRONO-LUME KIT NO 395
SDP
Catalog No. NC9801766
Encompass
CHRONO-LUME kit includes 1xP/N 387 ATP standard. Cost per test based on typical dilution of whole blood. Greater dilutions require less CHRONO-LUME reagent.
Catalog No. NC9801766 Supplier Chrono Log Corporation Supplier No. 395
CHRONO-LUME kit includes 1xP/N 387 ATP standard. Cost per test based on typical dilution of whole blood. Greater dilutions require less CHRONO-LUME reagent.