Learn More
Chrono Log Corporation CHRONO-PAR Ristocetin, Stabilized Freeze-Dried Reagent, Quantitative Testing for von Willebrand Disease, 62.5 mg
SDP
Catalog No. NC0532215
Encompass
The CHRONO-PAR Ristocetin from CHRONO LOG CORPORATION is a platelet aggregation reagent designed for accurate detection of von Willebrand Disease and Bernard Soulier Syndrome. It offers a cost-effective solution for clinical and research laboratories with a yield of 62 to 125 tests per vial, using a stabilized freeze-dried formulation. Compatible with both whole blood and PRP samples, it ensures reliable results in platelet function testing.
Catalog No. NC0532215 Supplier Chrono Log Corporation Supplier No. 396
The CHRONO-PAR Ristocetin from CHRONO LOG CORPORATION is a platelet aggregation reagent designed for accurate detection of von Willebrand Disease and Bernard Soulier Syndrome. It offers a cost-effective solution for clinical and research laboratories with a yield of 62 to 125 tests per vial, using a stabilized freeze-dried formulation. Compatible with both whole blood and PRP samples, it ensures reliable results in platelet function testing.