Learn More
Medchemexpress LLC HY-121107 500mg Medchemexpress, Chrysene CAS:218-01-9 Purity:>98%
SDP
Catalog No. 502080960
Encompass_Preferred
Medchemexpress, HY-121107 500mg Chrysene CAS:218-01-9 Chrysene is a high molecular weight (HMW), polycyclic aromatic hydrocarbon (PAH) known for its recalcitrance and carcinogenic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0960 Supplier Medchemexpress LLC Supplier No. HY121107500MG
Medchemexpress, HY-121107 500mg Chrysene CAS:218-01-9 Chrysene is a high molecular weight (HMW), polycyclic aromatic hydrocarbon (PAH) known for its recalcitrance and carcinogenic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.