Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysene 98%, Thermo Scientific™

Catalog No. AC224145000
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC224145000 500 mg
AC224140050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224145000 Supplier Thermo Scientific Chemicals Supplier No. 224145000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218-01-9
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
InChI Key WDECIBYCCFPHNR-UHFFFAOYSA-N
Synonym benzo a phenanthrene, 1,2-benzophenanthrene, 1,2-benzphenanthrene, 1,2,5,6-dibenzonaphthalene, benz a phenanthrene, chrysen, rcra waste number u050, rcra waste no. u050
PubChem CID 9171
ChEBI CHEBI:51687
IUPAC Name chrysene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43

Specifications

Melting Point 251°C to 255°C
Color Beige to White or Green-Yellow
Infrared Spectrum Authentic
Quantity 500 mg
Beilstein 05, 718
Merck Index 15, 2259
Formula Weight 228.29
Percent Purity ≥97.5%
Physical Form Powder or Flakes
Chemical Name or Material Chrysene, 98%
Hazard Category GHS Signal Word: Danger
Hazard Statement MUTAGEN
EINECSNumber 205-923-4
RTECSNumber GC0700000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.