Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysene, MilliporeSigma™ Supelco™

Catalog No. 111019030 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9030 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9030 Supplier MilliporeSigma Supelco Supplier No. 35754100MG
Add to Cart
Add to Cart

Chrysene is a symmetrical polycyclic aromatic hydrocarbon (PAH) with four condensed benzene rings. Incomplete combustion of fossil fuels and other industrial processes results in the production of chrysene which is known to be toxic, mutagenic and carcinogenic to humans.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 218-01-9
Physical Form Neat
Linear Formula C18H12
MDL Number MFCD00003698
Melting Point 252°C to 254°C
Boiling Point 448°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 228.29
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.