Learn More
Medchemexpress LLC Chryseno[2,3-d]dioxole-2,7(4aH,5H)-dione, 4b,6,6a,7a,10a,11,11a,11b,12,13-decahydro-5-hydroxy- | 1040085-99-1 | 98.6% | 430.53 g/mol | C25H34O6 | 5 MG
SDP
Catalog No. 5000215639
Encompass_Preferred
Budesonide impurity C is an analytical reference material used for impurity profiling, method validation, and quality-control testing in pharmaceutical analysis. The substance is characterized with a reported purity and supporting documentation to enable reliable use in laboratory assays.
  • High purity (98.6%) for reliable analytical results.
  • Suitable for impurity identification and method validation.
  • Supplied with certificate of analysis and datasheet.
  • Chemical formula C25H34O6 and molecular weight 430.53 g/mol.
  • Small quantity format appropriate for analytical testing.
Catalog No. 50-002-15639 Supplier Medchemexpress LLC Supplier No. HY1000875MG
Budesonide impurity C is an analytical reference material used for impurity profiling, method validation, and quality-control testing in pharmaceutical analysis. The substance is characterized with a reported purity and supporting documentation to enable reliable use in laboratory assays.
  • High purity (98.6%) for reliable analytical results.
  • Suitable for impurity identification and method validation.
  • Supplied with certificate of analysis and datasheet.
  • Chemical formula C25H34O6 and molecular weight 430.53 g/mol.
  • Small quantity format appropriate for analytical testing.