Learn More
Apexbio Technology LLC Chrysin, 1g Cas# 480-40-0
SDP
Supplier:  Apexbio Technology LLC N13361000
Encompass_Preferred
Chrysin is an antioxidant flavonoid. Chrysin displays anti-inflammatory and antitumor properties. Chrysin inhibits HIF-1 (hypoxia-inducible factor-1) and induces apoptosis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8566