Learn More
Apexbio Technology LLC Chrysin 480-40-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063744
Encompass_Preferred
Chrysin a naturally occurring flavonoid compound exhibits antioxidant activity and has been extensively studied in biomedical research contexts Primarily recognized for its capacity to modulate inflammatory signaling pathways and tumor-related cellular processes chrysin specifically acts through the inhibition of hypoxia-inducible factor-1 alpha (HIF-1 ) a critical regulator in cellular response to hypoxia Additionally chrysin promotes cell apoptosis indicating its potential role in antitumor protocols In experimental settings it serves as a biological tool to explore mechanisms underlying inflammation cancer progression and oxidative stress-mediated pathologies
Catalog No. 50-000-63744 Supplier Apexbio Technology LLC Supplier No. N13365.1
Chrysin a naturally occurring flavonoid compound exhibits antioxidant activity and has been extensively studied in biomedical research contexts Primarily recognized for its capacity to modulate inflammatory signaling pathways and tumor-related cellular processes chrysin specifically acts through the inhibition of hypoxia-inducible factor-1 alpha (HIF-1 ) a critical regulator in cellular response to hypoxia Additionally chrysin promotes cell apoptosis indicating its potential role in antitumor protocols In experimental settings it serves as a biological tool to explore mechanisms underlying inflammation cancer progression and oxidative stress-mediated pathologies