Learn More
Medchemexpress LLC Chrysin | 480-40-0 | MFCD00006834 | 100.0% | C15H10O4 | 10MG
SDP
Catalog No. 5000259388
Encompass_Preferred
Chrysin (Standard) is an analytical reference standard of chrysin (5,7-dihydroxyflavone) intended for research and analytical applications. It is used for compound identification, purity assessment, and method development in studies involving flavonoids and estrogen-related assays.
  • High purity: 99.96%.
  • Molecular formula: C15H10O4.
  • Intended for analytical and research applications.
  • Useful for compound identification and method development.
  • Available in multiple pack sizes, including 10 mg.
Catalog No. 50-002-59388 Supplier Medchemexpress LLC Supplier No. HY14589R10MG
Chrysin (Standard) is an analytical reference standard of chrysin (5,7-dihydroxyflavone) intended for research and analytical applications. It is used for compound identification, purity assessment, and method development in studies involving flavonoids and estrogen-related assays.
  • High purity: 99.96%.
  • Molecular formula: C15H10O4.
  • Intended for analytical and research applications.
  • Useful for compound identification and method development.
  • Available in multiple pack sizes, including 10 mg.