Learn More
Medchemexpress LLC Chrysin 6-C-glucoside 8-C-arabinoside | 185145-34-0 | 98.6% | 10 MG
SDP
Catalog No. 5000234177
Encompass_Preferred
Chrysin 6-C-glucoside 8-C-arabinoside is a flavonoid glycoside used in pharmacological research; it has been reported to inhibit calcitonin gene-related peptide (CGRP) release and to block TRPV1 channel activation, supporting anti-migraine and TRPV1-focused studies.
  • Reported inhibitor of CGRP release.
  • Blocks TRPV1 channel activation.
  • High purity suitable for in vitro pharmacology studies.
  • Molecular formula C26H28O13 and molecular weight 548.49 g/mol.
  • Available in small-mass packages for assay development.
Catalog No. 50-002-34177 Supplier Medchemexpress LLC Supplier No. HY12029410MG
Chrysin 6-C-glucoside 8-C-arabinoside is a flavonoid glycoside used in pharmacological research; it has been reported to inhibit calcitonin gene-related peptide (CGRP) release and to block TRPV1 channel activation, supporting anti-migraine and TRPV1-focused studies.
  • Reported inhibitor of CGRP release.
  • Blocks TRPV1 channel activation.
  • High purity suitable for in vitro pharmacology studies.
  • Molecular formula C26H28O13 and molecular weight 548.49 g/mol.
  • Available in small-mass packages for assay development.