Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chrysin 6-C-glucoside 8-C-arabinoside | 185145-34-0 | 98.6% | 5 MG
SDP

Catalog No. 5000234288
Encompass_Preferred

Chrysin 6-C-glucoside 8-C-arabinoside is a flavonoid glycoside used in preclinical research. It has been reported to inhibit calcitonin gene-related peptide (CGRP) release and to block activation of the TRPV1 channel, making it useful in studies related to migraine mechanisms and neuropharmacology.

  • Flavonoid glycoside that inhibits CGRP release and TRPV1 activation.
  • Used in anti-migraine and neuropharmacology research.
  • Molecular formula C26H28O13; molecular weight 548.49 g/mol.
  • Purity typically around 98.6% as supplied.
  • Available in small research packs (5 mg, 10 mg, 50 mg, 100 mg).
  • Store protected from light at 4°C; in solution, store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-34288 Supplier Medchemexpress LLC Supplier No. HY1202945MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chrysin 6-C-glucoside 8-C-arabinoside is a flavonoid glycoside used in preclinical research. It has been reported to inhibit calcitonin gene-related peptide (CGRP) release and to block activation of the TRPV1 channel, making it useful in studies related to migraine mechanisms and neuropharmacology.

  • Flavonoid glycoside that inhibits CGRP release and TRPV1 activation.
  • Used in anti-migraine and neuropharmacology research.
  • Molecular formula C26H28O13; molecular weight 548.49 g/mol.
  • Purity typically around 98.6% as supplied.
  • Available in small research packs (5 mg, 10 mg, 50 mg, 100 mg).
  • Store protected from light at 4°C; in solution, store at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.