Learn More
Cayman Chemical Chrysin 7-Glucuronide, Purity: ≥98%, CAS: 35775-49-6, 10 mg
SDP
Catalog No. 502913131
Encompass_Preferred
A flavonoid glucuronide with diverse biological activities and an active metabolite of chrysin; inhibits α-glucosidase, α-amylase, and neuraminidase (IC50s = 612, 980, and 428 µg/ml, respectively); inhibits OATP1A2, OATP1B1, OATP1B3, OATP2B1, BCRP, and MRP2 (IC50s = 24.1, 4.4, 14.3, 0.3, 19.8, and 11.2 µM, respectively)
Catalog No. 50-291-3131 Supplier Cayman Chemical Supplier No. 3503410 MG
A flavonoid glucuronide with diverse biological activities and an active metabolite of chrysin; inhibits α-glucosidase, α-amylase, and neuraminidase (IC50s = 612, 980, and 428 µg/ml, respectively); inhibits OATP1A2, OATP1B1, OATP1B3, OATP2B1, BCRP, and MRP2 (IC50s = 24.1, 4.4, 14.3, 0.3, 19.8, and 11.2 µM, respectively)