Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chrysin-7-O-glucuronide | 35775-49-6 | 1 MG
SDP

Catalog No. 5000420398
Encompass_Preferred

Chrysin-7-O-glucuronide is a flavonoid extracted from *Scutellaria baicalensis* that exhibits antioxidant activity. It is intended for research use only.

  • Purity: 99.47%
  • Source: Flavonoid extracted from *Scutellaria baicalensis*.
  • Activity: Antioxidant activity.
  • Appearance: Solid, off-white to yellow color.
  • Solubility (in vitro): 50 mg/mL (116.18 mM) in DMSO.
  • Structure classification: Flavonoids, flavones, phenols, monophenols.
  • Initial source: Plants (Leguminosae, Bignoniaceae, Labiatae).

Catalog No. 50-004-20398 Supplier Medchemexpress LLC Supplier No. HYN23761MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chrysin-7-O-glucuronide is a flavonoid extracted from *Scutellaria baicalensis* that exhibits antioxidant activity. It is intended for research use only.

  • Purity: 99.47%
  • Source: Flavonoid extracted from *Scutellaria baicalensis*.
  • Activity: Antioxidant activity.
  • Appearance: Solid, off-white to yellow color.
  • Solubility (in vitro): 50 mg/mL (116.18 mM) in DMSO.
  • Structure classification: Flavonoids, flavones, phenols, monophenols.
  • Initial source: Plants (Leguminosae, Bignoniaceae, Labiatae).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.