Learn More
Medchemexpress LLC Chrysin-7-O-glucuronide | 35775-49-6 | 1 ML
SDP
Catalog No. 5000421020
Encompass_Preferred
Chrysin-7-O-glucuronide is a flavonoid extracted from *Scutellaria baicalensis*, possessing antioxidant activity. It inhibits α-glucosidase and α-amylase, suppresses NF-κB signaling activity, scavenges free radicals, acts as a tight junction protector, and mitigates intestinal mucosal barrier injury. This compound is suitable for research related to type 2 diabetes and severe acute pancreatitis-induced intestinal barrier injury.
  • Inhibits alpha-glucosidase and alpha-amylase
  • Suppresses NF-κB signaling activity
  • Scavenges free radicals
  • Acts as a tight junction protector
  • Mitigates intestinal mucosal barrier injury
  • Restores ZO-1 expression and tight junction structure
  • Ameliorates SAP-induced intestinal mucosal barrier injury in rats
Catalog No. 50-004-21020 Supplier Medchemexpress LLC Supplier No. HYN237610MM/1ML
Chrysin-7-O-glucuronide is a flavonoid extracted from *Scutellaria baicalensis*, possessing antioxidant activity. It inhibits α-glucosidase and α-amylase, suppresses NF-κB signaling activity, scavenges free radicals, acts as a tight junction protector, and mitigates intestinal mucosal barrier injury. This compound is suitable for research related to type 2 diabetes and severe acute pancreatitis-induced intestinal barrier injury.
  • Inhibits alpha-glucosidase and alpha-amylase
  • Suppresses NF-κB signaling activity
  • Scavenges free radicals
  • Acts as a tight junction protector
  • Mitigates intestinal mucosal barrier injury
  • Restores ZO-1 expression and tight junction structure
  • Ameliorates SAP-induced intestinal mucosal barrier injury in rats