Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014267 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4267 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4267 Supplier MilliporeSigma Supelco Supplier No. 9508250MG
Add to Cart
Add to Cart

Chrysin is a naturally occurring mono-flavonoid , which is present in plant cells involved in photosynthesis. It may possess a broad spectrum of pharmacological properties like antioxidant, anti-viral, anti-inflammatory, anxiolytic and vasodilatory effects.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 480-40-0
Physical Form Neat
Linear Formula C15H10O4
MDL Number MFCD00006834
Melting Point 284°C to 286°C
Synonym 5,7-Dihydroxyflavone
Type Analytical Standard
Quantity 50 mg
Formula Weight 254.24
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.