Learn More
Abcam Chrysin, Antio x idant flavonoid, 1G
Catalog No. 502706777
Encompass_Preferred
MW 254.24 Da, Purity >99%. Antioxidant flavonoid. Anti-inflammatory and antioxidant properties, also able to induce apoptosis. Potent inhibitor of hypoxia-inducible factor 1α (HIF-1α).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6777 Supplier Abcam Supplier No. AB1412301G
MW 254.24 Da, Purity >99%. Antioxidant flavonoid. Anti-inflammatory and antioxidant properties, also able to induce apoptosis. Potent inhibitor of hypoxia-inducible factor 1α (HIF-1α).
The product is subject to the following: Abcam Restricted Use Statement