Learn More
Medchemexpress LLC Chrysin (Standard) | 480-40-0 | 100.0% | C₁₅H₁₀O₄ | 25 MG
SDP
Catalog No. 5000353052
Encompass_Preferred
Chrysin (Standard) is an analytical standard for Chrysin, primarily intended for research and analytical applications. Known as an estrogen blocker, it serves as a reference standard in various qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for Chrysin
  • Intended for research and analytical applications
  • Known as an estrogen blocker
  • Used as a reference standard in qualitative, quantitative, and methodological research
  • Suitable for techniques including HPLC, GC, and MS
Catalog No. 50-003-53052 Supplier Medchemexpress LLC Supplier No. 25MGHY14589R
Chrysin (Standard) is an analytical standard for Chrysin, primarily intended for research and analytical applications. Known as an estrogen blocker, it serves as a reference standard in various qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for Chrysin
  • Intended for research and analytical applications
  • Known as an estrogen blocker
  • Used as a reference standard in qualitative, quantitative, and methodological research
  • Suitable for techniques including HPLC, GC, and MS