Learn More
Medchemexpress LLC Chrysin (Standard) | 480-40-0 | 100.0% | C₁₅H₁₀O₄ | 50 MG
SDP
Catalog No. 5000354066
Encompass_Preferred
Chrysin (Standard) is the analytical standard of Chrysin, intended for research and analytical applications. It is known as one of the most well-known estrogen blockers.
  • Analytical standard grade
  • Supplied for assay reference
  • Commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-54066 Supplier Medchemexpress LLC Supplier No. HY14589R50MG
Chrysin (Standard) is the analytical standard of Chrysin, intended for research and analytical applications. It is known as one of the most well-known estrogen blockers.
  • Analytical standard grade
  • Supplied for assay reference
  • Commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.