Learn More
Medchemexpress LLC Chrysin (Standard) | 480-40-0 | 99.96% | C₁₅H₁₀O₄ | 100 MG
SDP
Catalog No. 5000354341
Encompass_Preferred
Chrysin (Standard), also known as 5,7-Dihydroxyflavone, is a high-purity analytical standard (99.96%) with the chemical formula C₁₅H₁₀O₄ and CAS number 480-40-0. This flavonoid is primarily utilized as a reference standard in various research and analytical applications, including qualitative, quantitative, and methodological experiments using techniques such as HPLC, GC, and MS.
  • Used as an analytical standard
  • Ideal for qualitative, quantitative, and methodological research experiments
  • Suitable for techniques like HPLC, GC, and MS
  • Functions as a reference standard for various assays
  • A naturally occurring flavonoid and flavone
  • Found in plants and as an endogenous metabolite in human gut microbiota
  • Known as an estrogen blocker, useful in estrogen receptor and ERR inhibition research
Catalog No. 50-003-54341 Supplier Medchemexpress LLC Supplier No. HY14589R100MG
Chrysin (Standard), also known as 5,7-Dihydroxyflavone, is a high-purity analytical standard (99.96%) with the chemical formula C₁₅H₁₀O₄ and CAS number 480-40-0. This flavonoid is primarily utilized as a reference standard in various research and analytical applications, including qualitative, quantitative, and methodological experiments using techniques such as HPLC, GC, and MS.
  • Used as an analytical standard
  • Ideal for qualitative, quantitative, and methodological research experiments
  • Suitable for techniques like HPLC, GC, and MS
  • Functions as a reference standard for various assays
  • A naturally occurring flavonoid and flavone
  • Found in plants and as an endogenous metabolite in human gut microbiota
  • Known as an estrogen blocker, useful in estrogen receptor and ERR inhibition research