Learn More
Cayman Chemical Linarin, Purity: ≥95%, CAS: 480-36-4, 50 mg
SDP
Catalog No. 502939234
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; potentiates TRAIL-induced cytotoxicity and apoptosis in U87MG human glioma cells; potentiates TRAIL-induced tumor cell apoptosis and reduction of tumor growth in a U87MG mouse xenograft model; reduces pulmonary platelet count, edema, and macrophage, polymorphonuclear leukocyte, and lymphocyte infiltration, as well as inhibits TXNIP/NLRP3, MAPK, and NF-κB signaling in a mouse model of LPS-induced acute lung injury from 12.5-50 mg/kg; inhibits AChE and induces dyskinesia recovery in a zebrafish model of Alzheimer's disease in tank water at 50 μg/ml
Catalog No. 50-293-9234 Supplier Cayman Chemical Supplier No. 2690050 MG
A flavonoid glycoside with diverse biological activities; potentiates TRAIL-induced cytotoxicity and apoptosis in U87MG human glioma cells; potentiates TRAIL-induced tumor cell apoptosis and reduction of tumor growth in a U87MG mouse xenograft model; reduces pulmonary platelet count, edema, and macrophage, polymorphonuclear leukocyte, and lymphocyte infiltration, as well as inhibits TXNIP/NLRP3, MAPK, and NF-κB signaling in a mouse model of LPS-induced acute lung injury from 12.5-50 mg/kg; inhibits AChE and induces dyskinesia recovery in a zebrafish model of Alzheimer's disease in tank water at 50 μg/ml