Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysoidine G, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018253 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8253 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8253 Supplier MilliporeSigma Supelco Supplier No. 80028100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 532-82-1
Physical Form Neat
Linear Formula C12H12N4 · HCl
MDL Number MFCD00012976
Melting Point 235°C
Synonym 4-Phenylazo-m-phenylenediamine monohydrochloride; Basic Orange 2; Chrysoidine Y
Type Analytical Standard
Quantity 100 mg
Formula Weight 248.71
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.