Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chrysoidine G (Solvent Orange 3 hydrochloride) | 532-82-1 | 250 MG
SDP

Catalog No. 5000414347
Encompass_Preferred

Chrysoidine G (Solvent Orange 3 hydrochloride) is an industrial azoic dye (cationic dye) used for the construction of most textile dyestuffs and in synthetic industrial compounds. Its concentration can be determined by UV-Vis spectroscopy. This product is for research use only and not for sale to patients.

  • Molecular weight: 248.71
  • Formula: C12H13ClN4
  • Appearance: Solid
  • Color: Brown to black
  • CAS number: 532-82-1

Catalog No. 50-004-14347 Supplier Medchemexpress LLC Supplier No. 250MGHYD0303A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chrysoidine G (Solvent Orange 3 hydrochloride) is an industrial azoic dye (cationic dye) used for the construction of most textile dyestuffs and in synthetic industrial compounds. Its concentration can be determined by UV-Vis spectroscopy. This product is for research use only and not for sale to patients.

  • Molecular weight: 248.71
  • Formula: C12H13ClN4
  • Appearance: Solid
  • Color: Brown to black
  • CAS number: 532-82-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.