Learn More
AdipoGen Chrysomycin A
SDP
Supplier:  AdipoGen BVT0099C250
Encompass_Preferred
Chemical. CAS 82196-88-1. Formula C28H28O9. MW 508.5. Isolated from Streptomyces sp. Antibiotic. Topoisomerase II inhibitor. Mediates a unique cross-linking reaction between DNA and histidine H3. Anti-neoplastic. Photosensitizing compound. Antitumor compound against human cell lines.
Catalog No. 50-256-2099