Learn More
AdipoGen Chrysomycin B
SDP
Supplier:  AdipoGen BVT0100M001
Encompass_Preferred
Chemical. CAS 83852-56-6. Formula C27H28O9. MW 496.5. Isolated from Streptomyces sp. Antibiotic. Topoisomerase II inhibitor. Mediates a unique cross-linking reaction between DNA and histidine H3. Anti-neoplastic. Photosensitizing compound. Antitumor compound against human cell lines. Less potent than chrysomycin A Prod. No. BVT-0099.
Catalog No. 50-256-2096