Learn More
Medchemexpress LLC Chrysoobtusin | 70588-06-6 | MFCD22124993 | 99.5% | 358.34 g/mol | C19H18O7 | 5 MG
SDP
Catalog No. 5000262545
Encompass_Preferred
Chrysoobtusin is an anthraquinone derivative isolated from Semen Cassiae, provided as a research-grade small molecule for laboratory studies. It is commonly used in biochemical and pharmacological research related to antioxidant, anti-inflammatory, and hepatoprotective activities.
  • Anthraquinone derivative isolated from Semen Cassiae.
  • High purity suitable for analytical and biological research.
  • Available in small-mass quantities for assay development and screening.
  • Identified by CAS number 70588-06-6 for unambiguous reference.
  • Molecular formula C19H18O7; molecular weight 358.34 g/mol.
Catalog No. 50-002-62545 Supplier Medchemexpress LLC Supplier No. HY1338605MG
Chrysoobtusin is an anthraquinone derivative isolated from Semen Cassiae, provided as a research-grade small molecule for laboratory studies. It is commonly used in biochemical and pharmacological research related to antioxidant, anti-inflammatory, and hepatoprotective activities.
  • Anthraquinone derivative isolated from Semen Cassiae.
  • High purity suitable for analytical and biological research.
  • Available in small-mass quantities for assay development and screening.
  • Identified by CAS number 70588-06-6 for unambiguous reference.
  • Molecular formula C19H18O7; molecular weight 358.34 g/mol.